2,6-Dichloro-4-(trifluoromethylsulphanyl)aniline 95%

Lieferant: Apollo Scientific

Synonyme: 2,6-Dichloro-4-(trifluoromethylthio)aniline

PC52455-1G PC52455-250MG
APOSPC52455-1GEA 487 EUR
APOSPC52455-1G APOSPC52455-250MG
2,6-Dichloro-4-(trifluoromethylsulphanyl)aniline 95%
2,6-Dichloro-4-(trifluoromethylsulphanyl)aniline
Formel: C₇H₄Cl₂F₃NS
Molecular Weight: 262,08 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04972800
CAS-Nummer: 99479-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR