2,3-Difluoro-5-ethoxytoluene 97%

Lieferant: Apollo Scientific

Synonyme: 5-Ethoxy-1,2-difluoro-3-methylbenzene

PC52479-1G PC52479-5G
APOSPC52479-1GEA 132 EUR
APOSPC52479-1G APOSPC52479-5G
2,3-Difluoro-5-ethoxytoluene 97%
2,3-Difluoro-5-ethoxytoluene
Formel: C₉H₁₀F₂O
Molecular Weight: 172.172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28788825
CAS-Nummer: 1803790-28-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR