2,6-Dibromo-4-(pentafluorosulphur)aniline 98%

Lieferant: Apollo Scientific

PC52481-1G PC52481-5G
APOSPC52481-1GEA 290 EUR
APOSPC52481-1G APOSPC52481-5G
2,6-Dibromo-4-(pentafluorosulphur)aniline 98%
2,6-Dibromo-4-(pentafluorosulphur)aniline
Formel: C₆H₄Br₂F₅NS
Molecular Weight: 376.968 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32691735
CAS-Nummer: 149757-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR