3-Bromo-2-hydroxy-5-(pentafluorothio)benzaldehyde 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Bromo-5-formyl-4-hydroxyphenylsulphur pentafluoride

PC5253-1G PC5253-250MG
APOSPC5253-1GEA 509 EUR
APOSPC5253-1G APOSPC5253-250MG
3-Bromo-2-hydroxy-5-(pentafluorothio)benzaldehyde 95+%
3-Bromo-2-hydroxy-5-(pentafluorothio)benzaldehyde
Formel: C₇H₄BrF₅O₂S
Molecular Weight: 327,066 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501045
CAS-Nummer: 1208718-46-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR