4-Bromo-2-chloro-3-fluorobenzonitrile 95%

Lieferant: Apollo Scientific

PC52716-1G PC52716-250MG PC52716-5G
APOSPC52716-1GEA 328 EUR
APOSPC52716-1G APOSPC52716-250MG APOSPC52716-5G
4-Bromo-2-chloro-3-fluorobenzonitrile 95%
4-Bromo-2-chloro-3-fluorobenzonitrile
Formel: C₇H₂BrClFN
Molecular Weight: 234.453 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23707414
CAS-Nummer: 1427439-32-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR