2-Iodo-3-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carboxy-2-iodo-α,α,α-trifluorotoluene, 3-Carboxy-2-iodobenzotrifluoride

PC5275-1G PC5275-5G
APOSPC5275-1GEA 165 EUR
APOSPC5275-1G APOSPC5275-5G
2-Iodo-3-(trifluoromethyl)benzoic acid
2-Iodo-3-(trifluoromethyl)benzoic acid
Formel: C₈H₄F₃IO₂
Molecular Weight: 316,016 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08059374
CAS-Nummer: 766473-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR