2,6-Difluoro-D-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: (R)-2-Amino-3-(2,6-difluorophenyl)propanoic acid

PC530004-1G PC530004-250MG
APOSPC530004-1GEA 351 EUR
APOSPC530004-1G APOSPC530004-250MG
2,6-Difluoro-D-phenylalanine 97%
2,6-Difluoro-D-phenylalanine
Formel: C₉H₉F₂NO₂
Molecular Weight: 201.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07372005
CAS-Nummer: 266360-62-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR