2,4-Difluoro-L-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Amino-3-(2,4-difluorophenyl)propanoic acid

PC530005-1G
APOSPC530005-1GEA 69.6 EUR
APOSPC530005-1G
2,4-Difluoro-L-phenylalanine 97%
2,4-Difluoro-L-phenylalanine
Formel: C₉H₉F₂NO₂
Molecular Weight: 201,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03844652
CAS-Nummer: 31105-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR