4-Fluoro-3-(trifluoromethoxy)benzyl alcohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[4-Fluoro-3-(trifluoromethoxy)phenyl]methanol
PC53001-1G
PC53001-5G
PC53001-10G
PC53001-25G
APOSPC53001-1GEA
249
EUR
APOSPC53001-1G
APOSPC53001-5G
APOSPC53001-10G
APOSPC53001-25G
4-Fluoro-3-(trifluoromethoxy)benzyl alcohol 98%
4-Fluoro-3-(trifluoromethoxy)benzyl alcohol
Formel:
C₈H₆F₄O₂ Molecular Weight: 210,126 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06660255 CAS-Nummer: 86256-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...