Fmoc-3-Fluoro-D-b-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-fluorophenyl)propanoic acid

PC530013-1G
APOSPC530013-1GEA 150 EUR
APOSPC530013-1G
Fmoc-3-Fluoro-D-b-phenylalanine 97%
Fmoc-3-Fluoro-D-b-phenylalanine
Formel: C₂₄H₂₀FNO₄
Molecular Weight: 405,418 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03428032
CAS-Nummer: 511272-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR