(R)-3-Amino-4-(2,4,5-trifluorophenyl)butyric acid 97%

Lieferant: Apollo Scientific

Synonyme: (R)-3-Amino-4-(2,4,5-trifluorophenyl)butanoic acid

PC530016-1G PC530016-5G
APOSPC530016-1GEA 59.6 EUR
APOSPC530016-1G APOSPC530016-5G
(R)-3-Amino-4-(2,4,5-trifluorophenyl)butyric acid 97%
(R)-3-Amino-4-(2,4,5-trifluorophenyl)butyric acid
Formel: C₁₀H₁₀F₃NO₂
Molecular Weight: 233.187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07363507
CAS-Nummer: 936630-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR