4'-Fluoro-biphenyl-3-acetic acid 97%
Lieferant: Apollo Scientific
PC530019-1G
PC530019-250MG
APOSPC530019-1GEA
654
EUR
APOSPC530019-1G
APOSPC530019-250MG
4'-Fluoro-biphenyl-3-acetic acid 97%
4'-Fluoro-biphenyl-3-acetic acid
Formel:
C₁₄H₁₁FO₂ Molecular Weight: 230,234 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03426493 CAS-Nummer: 327107-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...