(2Z)-3-(Diethylamino)-2-fluoroprop-2-enal 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2Z)-3-(Diethylamino)-2-fluoroacrolein, (2Z)-3-(Diethylamino)-2-fluoroacrylaldehyde

PC53002-1G PC53002-5G
APOSPC53002-1GEA 139 EUR
APOSPC53002-1G APOSPC53002-5G
(2Z)-3-(Diethylamino)-2-fluoroprop-2-enal 95%
(2Z)-3-(Diethylamino)-2-fluoroprop-2-enal
Formel: C₇H₁₂FNO
Molecular Weight: 145,175 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD18633001
CAS-Nummer: 152873-64-8
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR