4-Trifluoromethylphenethyl bromide 97%
Lieferant: Apollo Scientific
PC530024-1G
PC530024-5G
APOSPC530024-1GEA
0
EUR
APOSPC530024-1G
APOSPC530024-5G
4-Trifluoromethylphenethyl bromide 97%
4-Trifluoromethylphenethyl bromide
Formel:
C₉H₈BrF₃ Dichte: 1,499 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD10567022 CAS-Nummer: 130365-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...