5-Fluoro-L-tryptophan 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Amino-3-(5-fluoro-1H-indol-3-yl)propanoic acid, H-Trp(5-F)-OH

PC530031-1G PC530031-250MG PC530031-500MG
APOSPC530031-1GEA 870 EUR
APOSPC530031-1G APOSPC530031-250MG APOSPC530031-500MG
5-Fluoro-L-tryptophan 97%
5-Fluoro-L-tryptophan
Formel: C₁₁H₁₁FN₂O₂
Molecular Weight: 222.216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066470
CAS-Nummer: 16626-02-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR