7-Fluoro-L-tryptophan 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-Amino-3-(7-fluoro-1H-indol-3-yl)propanoic acid, H-Trp(7-F)-OH

PC530032-100MG PC530032-1G PC530032-250MG
APOSPC530032-100MGEA 183 EUR
APOSPC530032-100MG APOSPC530032-1G APOSPC530032-250MG
7-Fluoro-L-tryptophan 95%
7-Fluoro-L-tryptophan
Formel: C₁₁H₁₁FN₂O₂
Molecular Weight: 222.216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264337
CAS-Nummer: 138514-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR