b-Amino-4-fluorobenzenepropanol 97%

Lieferant: Apollo Scientific

PC530045-1G PC530045-250MG PC530045-500MG
APOSPC530045-1GEA 783 EUR
APOSPC530045-1G APOSPC530045-250MG APOSPC530045-500MG
b-Amino-4-fluorobenzenepropanol 97%
b-Amino-4-fluorobenzenepropanol
Formel: C₉H₁₂FNO
Molecular Weight: 169,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01697331
CAS-Nummer: 35373-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR