(S)-b-Amino-4-fluorobenzenepropanol 97%

Lieferant: Apollo Scientific

PC530047-1G PC530047-250MG PC530047-500MG
APOSPC530047-1GEA 870 EUR
APOSPC530047-1G APOSPC530047-250MG APOSPC530047-500MG
(S)-b-Amino-4-fluorobenzenepropanol 97%
(S)-b-Amino-4-fluorobenzenepropanol
Formel: C₉H₁₂FNO
Molecular Weight: 169.196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17214637
CAS-Nummer: 200267-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR