(S)-b-Amino-3-(trifluoromethyl)benzenepropanol 97%

Lieferant: Apollo Scientific

PC530050-1G PC530050-250MG PC530050-500MG
APOSPC530050-1GEA 870 EUR
APOSPC530050-1G APOSPC530050-250MG APOSPC530050-500MG
(S)-b-Amino-3-(trifluoromethyl)benzenepropanol 97%
(S)-b-Amino-3-(trifluoromethyl)benzenepropanol
Formel: C₁₀H₁₂F₃NO
Molecular Weight: 219,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17214661
CAS-Nummer: 938462-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR