(S)-b-Amino-3-fluorobenzenepropanol 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Amino-3-(3-fluorophenyl)propan-1-ol

PC530051-1G PC530051-250MG
APOSPC530051-1GEA 579 EUR
APOSPC530051-1G APOSPC530051-250MG
(S)-b-Amino-3-fluorobenzenepropanol 97%
(S)-b-Amino-3-fluorobenzenepropanol
Formel: C₉H₁₂FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD17214664
CAS-Nummer: 938462-29-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR