3-Fluoro-4-[4-(Boc-amino)piperidino]aniline 97%

Lieferant: Apollo Scientific

PC530054-1G PC530054-250MG PC530054-500MG
APOSPC530054-1GEA 163 EUR
APOSPC530054-1G APOSPC530054-250MG APOSPC530054-500MG
3-Fluoro-4-[4-(Boc-amino)piperidino]aniline 97%
3-Fluoro-4-[4-(Boc-amino)piperidino]aniline
Formel: C₁₆H₂₄FN₃O₂
Molecular Weight: 309,379 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23701381
CAS-Nummer: 1000053-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR