Methyl 2,2,2-trifluoroethyl carbonate 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC53009-1G
PC53009-5G
PC53009-25G
APOSPC53009-1GEA
21.9
EUR
APOSPC53009-1G
APOSPC53009-5G
APOSPC53009-25G
Methyl 2,2,2-trifluoroethyl carbonate 95%
Methyl 2,2,2-trifluoroethyl carbonate
Formel:
C₄H₅F₃O₃ Siedepunkt: 57…59 °C ( 120 mm Hg) Flammpunkt: 38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054295 CAS-Nummer: 156783-95-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...