3-Bromo-2-chloro-6-fluoroaniline, tech

Lieferant: Apollo Scientific
Achtung

PC53023-1G PC53023-5G
APOSPC53023-1GEA 200 EUR
APOSPC53023-1G APOSPC53023-5G
3-Bromo-2-chloro-6-fluoroaniline, tech
3-Bromo-2-chloro-6-fluoroaniline
Formel: C₆H₄BrClFN
Molecular Weight: 224.458 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28140550
CAS-Nummer: 1702023-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR