5-Fluoro-2-(2,2,2-trifluoroethoxy)aniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-β,β,β,4-tetrafluorophenetole

PC53030-1G PC53030-5G
APOSPC53030-1GEA 183 EUR
APOSPC53030-1G APOSPC53030-5G
5-Fluoro-2-(2,2,2-trifluoroethoxy)aniline
5-Fluoro-2-(2,2,2-trifluoroethoxy)aniline
Formel: C₈H₇F₄NO
Molecular Weight: 209.141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08699546
CAS-Nummer: 334929-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR