6-Fluoro-4-(trifluoromethyl)-1H-benzimidazole
Lieferant: Apollo Scientific
|
Achtung
|
PC53032-1G
PC53032-250MG
APOSPC53032-1GEA
406
EUR
APOSPC53032-1G
APOSPC53032-250MG
6-Fluoro-4-(trifluoromethyl)-1H-benzimidazole
6-Fluoro-4-(trifluoromethyl)-1H-benzimidazole
Formel:
C₈H₄F₄N₂ Molecular Weight: 204,124 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28155024 CAS-Nummer: 1360936-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...