4'-Nitro-2,2,2-trifluoroacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,2-Trifluoro-1-(4-nitrophenyl)ethanone

PC53033-1G PC53033-250MG PC53033-5G
APOSPC53033-1GEA 287 EUR
APOSPC53033-1G APOSPC53033-250MG APOSPC53033-5G
4'-Nitro-2,2,2-trifluoroacetophenone
4'-Nitro-2,2,2-trifluoroacetophenone
Formel: C₈H₄F₃NO₃
Molecular Weight: 219.117 g/mol
Schmelzpunkt: 42…47 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13173414
CAS-Nummer: 58808-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR