3-Fluorobicyclo[1.1.1]pentan-1-amine hydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC53046-100MG
PC53046-250MG
APOSPC53046-100MGEA
575
EUR
APOSPC53046-100MG
APOSPC53046-250MG
3-Fluorobicyclo[1.1.1]pentan-1-amine hydrochloride 95%
3-Fluorobicyclo[1.1.1]pentan-1-amine hydrochloride
Formel:
C₅H₈FN·ClH Molecular Weight: 137,583 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28963742 CAS-Nummer: 1826900-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...