Bromo(difluoro)acetic anhydride

Lieferant: Apollo Scientific
Gefahr

PC53049-25G PC53049-100G PC53049-5G
APOSPC53049-25GEA 114 EUR
APOSPC53049-25G APOSPC53049-100G APOSPC53049-5G
Bromo(difluoro)acetic anhydride
Bromo(difluoro)acetic anhydride
Formel: C₄Br₂F₄O₃
Siedepunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD24842567
CAS-Nummer: 7601-98-1
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR