Perfluoro(3-ethylpentane)

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,1,1,2,2,3,4,4,5,5,5-Undecafluoro-3-(1,1,2,2,2-pentafluoroethyl)pentane, Flutec PP70

PC53050-1G PC53050-5G
APOSPC53050-1GEA 94.8 EUR
APOSPC53050-1G APOSPC53050-5G
Perfluoro(3-ethylpentane)
Perfluoro(3-ethylpentane)
Formel: C₇F₁₆
Molecular Weight: 388.049 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28900770
CAS-Nummer: 2690-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR