4-(Trifluoroacetyl)phenacyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4'-(Bromoacetyl)-2,2,2-trifluoroacetophenone, 1-[4-(Bromoacetyl)phenyl]-2,2,2-trifluoroethanone

PC53051-1G PC53051-250MG PC53051-5G
APOSPC53051-1GEA 109 EUR
APOSPC53051-1G APOSPC53051-250MG APOSPC53051-5G
4-(Trifluoroacetyl)phenacyl bromide
4-(Trifluoroacetyl)phenacyl bromide
Formel: C₁₀H₆BrF₃O₂
Molecular Weight: 295.053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28963745
CAS-Nummer: 1980063-91-9
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR