17-Bromo-1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoroheptadecane
Lieferant: Apollo Scientific
|
Achtung
|
PC53054-1G
PC53054-250MG
APOSPC53054-1GEA
393
EUR
APOSPC53054-1G
APOSPC53054-250MG
17-Bromo-1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoroheptadecane
17-Bromo-1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoroheptadecane
Formel:
C₁₇H₂₂BrF₁₃ Molecular Weight: 553,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28963748 CAS-Nummer: 155401-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...