2-Bromo-5-(trifluoromethyl)pyrimidine 95+%

Lieferant: Apollo Scientific
Achtung

PC53065-250MG PC53065-1G
APOSPC53065-250MGEA 72.5 EUR
APOSPC53065-250MG APOSPC53065-1G
2-Bromo-5-(trifluoromethyl)pyrimidine 95+%
2-Bromo-5-(trifluoromethyl)pyrimidine
Formel: C₅H₂BrF₃N₂
Molecular Weight: 226,982 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD14702304
CAS-Nummer: 69034-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR