Potassium (3-acetylphenyl)trifluoroborate

Lieferant: Apollo Scientific
Achtung

PC53075-1G PC53075-10G
APOSPC53075-1GEA 29 EUR
APOSPC53075-1G APOSPC53075-10G
Potassium (3-acetylphenyl)trifluoroborate
Potassium (3-acetylphenyl)trifluoroborate
Formel: C₈H₇BF₃O·K
Molecular Weight: 226,045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368352
CAS-Nummer: 854906-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR