Perfluorobutyl bromide 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-1,1,2,2,3,3,4,4,4-nonafluorobutane

PC5309-5G PC5309-1G
APOSPC5309-5GEA 183 EUR
APOSPC5309-5G APOSPC5309-1G
Perfluorobutyl bromide 97%
Perfluorobutyl bromide
Formel: C₄BrF₉
Molecular Weight: 298,932 g/mol
Siedepunkt: 43…44 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082622
CAS-Nummer: 375-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR