2,4-Difluoro-5-iodoanisole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,5-Difluoro-2-iodo-4-methoxybenzene
PC53097-1G
PC53097-5G
APOSPC53097-1GEA
219
EUR
APOSPC53097-1G
APOSPC53097-5G
2,4-Difluoro-5-iodoanisole
2,4-Difluoro-5-iodoanisole
Formel:
C₇H₅F₂IO Molecular Weight: 270,015 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28986200 CAS-Nummer: 1936250-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...