4-(Difluoromethoxy)-3-fluorobenzaldehyde

Lieferant: Apollo Scientific
Achtung

PC53100-1G PC53100-25G PC53100-5G
APOSPC53100-1GEA 65.4 EUR
APOSPC53100-1G APOSPC53100-25G APOSPC53100-5G
4-(Difluoromethoxy)-3-fluorobenzaldehyde
4-(Difluoromethoxy)-3-fluorobenzaldehyde
Formel: C₈H₅F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11847153
CAS-Nummer: 1214379-56-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR