3-Bromo-5-fluoro-2-nitrophenol

Lieferant: Apollo Scientific
Achtung

PC53104-1G PC53104-250MG
APOSPC53104-1GEA 406 EUR
APOSPC53104-1G APOSPC53104-250MG
3-Bromo-5-fluoro-2-nitrophenol
3-Bromo-5-fluoro-2-nitrophenol
Formel: C₆H₃BrFNO₃
Molecular Weight: 235,995 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28740343
CAS-Nummer: 1807155-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR