5-Bromo-2,4-difluorophenol 95+%

Lieferant: Apollo Scientific
Achtung

PC53121-1G PC53121-250MG PC53121-5G
APOSPC53121-1GEA 105 EUR
APOSPC53121-1G APOSPC53121-250MG APOSPC53121-5G
5-Bromo-2,4-difluorophenol 95+%
5-Bromo-2,4-difluorophenol
Formel: C₆H₃BrF₂O
Molecular Weight: 208,988 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20231458
CAS-Nummer: 355423-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR