2,2-Difluoro-1,3-benzodioxole-5-acetic acid
Lieferant: Apollo Scientific
Synonyme:
2-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)acetic acid
PC53149-1G
PC53149-5G
APOSPC53149-1GEA
234
EUR
APOSPC53149-1G
APOSPC53149-5G
2,2-Difluoro-1,3-benzodioxole-5-acetic acid
2,2-Difluoro-1,3-benzodioxole-5-acetic acid
Formel:
C₉H₆F₂O₄ Molecular Weight: 216,138 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09925076 CAS-Nummer: 398156-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...