2,5-Difluoro-4-hydroxybenzonitrile

Lieferant: Apollo Scientific
Achtung

PC53157-1G PC53157-250MG
APOSPC53157-1GEA 295 EUR
APOSPC53157-1G APOSPC53157-250MG
2,5-Difluoro-4-hydroxybenzonitrile
2,5-Difluoro-4-hydroxybenzonitrile
Formel: C₇H₃F₂NO
Molecular Weight: 155.102 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258917
CAS-Nummer: 887267-05-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR