4-Fluoro-3-hydroxybenzaldehyde 98%

Lieferant: Apollo Scientific
Achtung

PC53163-1G PC53163-5G
APOSPC53163-1GEA 78.3 EUR
APOSPC53163-1G APOSPC53163-5G
4-Fluoro-3-hydroxybenzaldehyde 98%
4-Fluoro-3-hydroxybenzaldehyde
Formel: C₇H₅FO₂
Molecular Weight: 140,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11110190
CAS-Nummer: 103438-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR