(2R)-2-Amino-1,1,1-trifluoropropane 99%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2R)-1,1,1-Trifluoropropan-2-amine

PC53181-1G PC53181-250MG
APOSPC53181-1GEA 542 EUR
APOSPC53181-1G APOSPC53181-250MG
(2R)-2-Amino-1,1,1-trifluoropropane 99%
(2R)-2-Amino-1,1,1-trifluoropropane
Formel: C₃H₆F₃N
Molecular Weight: 113.082 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16817687
CAS-Nummer: 779303-24-1
UN: 2733
ADR: 3+8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR