2,6-Difluoro-3,5-dinitrotoluene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4-Difluoro-3-methyl-1,5-dinitrobenzene
PC53183-1G
PC53183-250MG
APOSPC53183-1GEA
509
EUR
APOSPC53183-1G
APOSPC53183-250MG
2,6-Difluoro-3,5-dinitrotoluene
2,6-Difluoro-3,5-dinitrotoluene
Formel:
C₇H₄F₂N₂O₄ Molecular Weight: 218,114 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16620286 CAS-Nummer: 112822-76-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...