Potassium (3,4,5-trifluorophenyl)trifluoroborate

Lieferant: Apollo Scientific
Achtung

PC53184-1G PC53184-5G
APOSPC53184-1GEA 183 EUR
APOSPC53184-1G APOSPC53184-5G
Potassium (3,4,5-trifluorophenyl)trifluoroborate
Potassium (3,4,5-trifluorophenyl)trifluoroborate
Formel: C₆H₂BF₆·K
Molecular Weight: 237,98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09993498
CAS-Nummer: 267006-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR