HFPO octamer alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: 1H,1H-Perfluoro(2,5,8,11,14,17-hexamethyl-3,6,9,12,15,18-oxatetracosan-1-ol), Monohydroxy perfluoropolyether

PC53216-1G PC53216-5G
APOSPC53216-1GEA 357 EUR
APOSPC53216-1G APOSPC53216-5G
HFPO octamer alcohol
HFPO octamer alcohol
Formel: C₂₄H₃F₄₇O₈
Molecular Weight: 1312.201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054285
CAS-Nummer: 1980064-28-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR