1-(4-Nitrophenyl)-2,2,2-trifluoroethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2,2-Trifluoro-1-(4-nitrophenyl)ethanol
PC53231-1G
PC53231-250MG
PC53231-5G
APOSPC53231-1GEA
289
EUR
APOSPC53231-1G
APOSPC53231-250MG
APOSPC53231-5G
1-(4-Nitrophenyl)-2,2,2-trifluoroethanol
1-(4-Nitrophenyl)-2,2,2-trifluoroethanol
Formel:
C₈H₆F₃NO₃ Molecular Weight: 221.133 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16615353 CAS-Nummer: 241127-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...