Methyl 3-fluoro-5-methylisonicotinate

Lieferant: Apollo Scientific
Achtung

PC53259-250MG PC53259-500MG PC53259-100MG
APOSPC53259-250MGEA 227 EUR
APOSPC53259-250MG APOSPC53259-500MG APOSPC53259-100MG
Methyl 3-fluoro-5-methylisonicotinate
Methyl 3-fluoro-5-methylisonicotinate
Formel: C₈H₈FNO₂
Molecular Weight: 169,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28798350
CAS-Nummer: 1803821-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR