Potassium 2-(trifluoromethyl)phenyltrifluoroborate

Lieferant: Apollo Scientific
Achtung

PC53272-1G PC53272-5G
APOSPC53272-1GEA 48 EUR
APOSPC53272-1G APOSPC53272-5G
Potassium 2-(trifluoromethyl)phenyltrifluoroborate
Potassium 2-(trifluoromethyl)phenyltrifluoroborate
Formel: C₇H₄BF₆·K
Molecular Weight: 252,006 g/mol
Schmelzpunkt: 175…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD15144778
CAS-Nummer: 30678-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR