((3S,4R)-4-(4-Fluorophenyl)piperidin-3-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
PC53318-1G
PC53318-250MG
APOSPC53318-1GEA
352
EUR
APOSPC53318-1G
APOSPC53318-250MG
((3S,4R)-4-(4-Fluorophenyl)piperidin-3-yl)methanol
((3S,4R)-4-(4-Fluorophenyl)piperidin-3-yl)methanol
Formel:
C₁₂H₁₆FNO Molecular Weight: 209,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03839921 CAS-Nummer: 125224-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...