Potassium (3-fluoro-4-formylphenyl)trifluoroborate
Lieferant: Apollo Scientific
|
Achtung
|
PC53344-1G
PC53344-5G
PC53344-25G
APOSPC53344-1GEA
58
EUR
APOSPC53344-1G
APOSPC53344-5G
APOSPC53344-25G
Potassium (3-fluoro-4-formylphenyl)trifluoroborate
Potassium (3-fluoro-4-formylphenyl)trifluoroborate
Formel:
C₇H₄BF₄O·K Molecular Weight: 230.009 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09993334 CAS-Nummer: 1451390-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...